Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gtpbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002454.1 | NP_001002454.1 | 9 - 10 | 973 | 87 | |
GenBank mRNA | BC076017.1 | - | 9 - 10 | 973 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |