Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp6v0a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020666.1 | NP_001018502.1 | 18 - 19 | 2230 | 74 | |
XM_009297243.2 | XP_009295518.1 | 18 - 19 | 2354 | 74 | ||
GenBank mRNA | BC095691.1 | - | 18 - 19 | 2230 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |