Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024665.1 | NP_001019836.1 | 2 - 3 | 723 | 128 | |
XM_005160883.4 | XP_005160940.1 | 2 - 3 | 128 | |||
GenBank mRNA | DQ060246.1 | - | 2 - 3 | 723 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |