Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: elnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048064.2 | NP_001041529.2 | 54 - 55 | 5796 | 114 | |
XM_017352793.2 | XP_017208282.1 | 53 - 54 | 6071 | 114 | ||
GenBank mRNA | DQ523563.1 | - | 54 - 55 | 5898 | 114 | |
MF118597.1 | - | 6 - 7 | 512 | 114 | ||
MF118598.1 | - | 6 - 7 | 455 | 114 | ||
MF118599.1 | - | 6 - 7 | 512 | 114 | ||
MF118600.1 | - | 6 - 7 | 512 | 114 | ||
MF118601.1 | - | 6 - 7 | 512 | 114 | ||
MF118602.1 | - | 6 - 7 | 455 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |