Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zp2l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089502.1 | NP_001082971.1 | 1365 | 97 | ||
GenBank mRNA | BC134168.1 | - | 1365 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio |