Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131113.1 | NP_571188.1 | 3 - 4 | 386 | 59 | |
XM_017353502.2 | XP_017208991.1 | 3 - 4 | 592 | 59 | ||
GenBank mRNA | BC162134.1 | - | 3 - 4 | 228 | 59 | |
BC162146.1 | - | 3 - 4 | 228 | 59 | ||
X68432.1 | - | 386 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |