Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: xpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200471.1 | NP_956765.1 | 4 - 5 | 476 | 115 | |
XM_005159814.4 | XP_005159871.1 | 4 - 5 | 520 | 115 | ||
XM_009306421.3 | XP_009304696.1 | 4 - 5 | 534 | 115 | ||
XM_009306424.3 | XP_009304699.1 | 4 - 5 | 518 | 115 | ||
GenBank mRNA | BC055179.1 | - | 4 - 5 | 476 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |