Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pafah1b1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201345.1 | NP_958502.1 | 4 - 5 | 507 | 110 | |
XM_005157575.4 | XP_005157632.1 | 4 - 5 | 500 | 110 | ||
GenBank mRNA | BC053205.1 | - | 4 - 5 | 507 | 110 | |
BC164856.1 | - | 3 - 4 | 229 | 110 | ||
DQ141217.1 | - | 3 - 4 | 288 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |