Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stt3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201458.2 | NP_958866.1 | 9 - 10 | 1065 | 77 | |
GenBank mRNA | BC046072.1 | - | 9 - 10 | 1066 | 77 | |
BC063234.1 | - | 9 - 10 | 1026 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |