Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alcamb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212634.3 | NP_997799.3 | 10 - 11 | 1349 | 68 | |
XM_005157504.4 | XP_005157561.1 | 10 - 11 | 1360 | 68 | ||
GenBank mRNA | AY050561.1 | - | 10 - 11 | 1305 | 68 | |
BC163458.1 | - | 10 - 11 | 1255 | 68 | ||
BC163486.1 | - | 10 - 11 | 1255 | 68 | ||
CU458904.1 | - | 10 - 11 | 1219 | 68 | ||
CU458936.1 | - | 10 - 11 | 1271 | 68 | ||
DQ279080.1 | - | 10 - 11 | 1312 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |