Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stx12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002051.1 | NP_001002051.1 | 3 - 4 | 295 | 72 | |
XM_009294127.3 | XP_009292402.1 | 3 - 4 | 350 | 72 | ||
XM_009294128.2 | XP_009292403.1 | 3 - 4 | 298 | 72 | ||
GenBank mRNA | BC071327.1 | - | 3 - 4 | 295 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |