Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcgf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007158.2 | NP_001007159.1 | 7 - 8 | 704 | 89 | |
XM_009307695.3 | XP_009305970.1 | 7 - 8 | 838 | 89 | ||
GenBank mRNA | BC134012.1 | - | 7 - 8 | 704 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |