Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ehmt1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030131.2 | NP_001025302.1 | 19 - 20 | 2637 | 69 | |
XM_005171778.4 | XP_005171835.1 | 20 - 21 | 2809 | 69 | ||
XM_005171779.4 | XP_005171836.1 | 20 - 21 | 2806 | 69 | ||
XM_021475966.1 | XP_021331641.1 | 18 - 19 | 2602 | 69 | ||
GenBank mRNA | BC115049.1 | - | 7 - 8 | 717 | 69 | |
BC133980.1 | - | 4 - 5 | 69 | |||
BC171681.1 | - | 19 - 20 | 2668 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |