Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcsk5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021476634.1 | XP_021332309.1 | 17 - 18 | 2502 | 71 | |
GenBank mRNA | DQ983232.1 | - | 17 - 18 | 2431 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Danio rerio Probe spans exons |