Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eomesa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131679.3 | NP_571754.3 | 1 - 2 | 840 | 93 | |
XM_005159319.4 | XP_005159376.1 | 2 - 3 | 1288 | 93 | ||
XM_005159321.4 | XP_005159378.1 | 2 - 3 | 1174 | 93 | ||
GenBank mRNA | AF287007.2 | - | 2 - 3 | 744 | 93 | |
AF329830.1 | - | 1 - 2 | 802 | 93 | ||
BC067719.1 | - | 1 - 2 | 840 | 93 | ||
BC154205.1 | - | 2 - 3 | 1063 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |