Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dixdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182884.1 | NP_878304.1 | 14 - 15 | 1720 | 67 | |
GenBank mRNA | AF502143.1 | - | 14 - 15 | 1720 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |