Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snapc1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200254.1 | NP_956548.1 | 3 - 4 | 571 | 142 | |
GenBank mRNA | BC048040.1 | - | 3 - 4 | 571 | 142 | |
BC164392.1 | - | 3 - 4 | 466 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |