Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: surf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201127.1 | NP_957421.1 | 5 - 6 | 673 | 66 | |
GenBank mRNA | BC056295.1 | - | 5 - 6 | 673 | 66 | |
BC066509.1 | - | 5 - 6 | 659 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |