Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdr2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001316894.1 | NP_001303823.1 | 5 - 6 | 751 | 65 | |
NM_205564.2 | NP_991127.2 | 4 - 5 | 965 | 65 | ||
XM_021469707.1 | XP_021325382.1 | 5 - 6 | 903 | 65 | ||
XM_021469708.1 | XP_021325383.1 | 5 - 6 | 769 | 65 | ||
GenBank mRNA | BC065979.1 | - | 4 - 5 | 941 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |