Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2x6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013565.2 | NP_001013583.1 | 2 - 3 | 368 | 63 | |
GenBank mRNA | BC078308.1 | - | 2 - 3 | 373 | 63 | |
BC090406.1 | - | 2 - 3 | 372 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |