Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: erc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001015065.1 | NP_001015065.1 | 2 - 3 | 1072 | 119 | |
XM_009298003.3 | XP_009296278.1 | 3 - 4 | 1417 | 119 | ||
XM_009298005.3 | XP_009296280.1 | 3 - 4 | 1418 | 119 | ||
XM_009298006.3 | XP_009296281.1 | 3 - 4 | 1416 | 119 | ||
XM_021470486.1 | XP_021326161.1 | 2 - 3 | 1147 | 119 | ||
XM_021470487.1 | XP_021326162.1 | 2 - 3 | 1066 | 119 | ||
GenBank mRNA | AY916778.1 | - | 2 - 3 | 1072 | 119 | |
BC171689.1 | - | 2 - 3 | 1147 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |