Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttc9b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044905.2 | NP_001038370.1 | 2 - 3 | 784 | 56 | |
GenBank mRNA | BC127585.1 | - | 2 - 3 | 784 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |