Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mc4r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173278.1 | NP_775385.1 | 1 | 549 | 79 | |
GenBank mRNA | BC163219.1 | - | 1 | 616 | 79 | |
BC163227.1 | - | 1 | 613 | 79 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |