Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: psma8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213166.1 | NP_998331.1 | 1 - 2 | 203 | 70 | |
GenBank mRNA | BC065608.1 | - | 1 - 2 | 203 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |