Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hccsa.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002498.2 | NP_001002498.2 | 6 - 7 | 697 | 62 | |
XM_005163321.4 | XP_005163378.1 | 5 - 6 | 585 | 62 | ||
GenBank mRNA | BC076303.1 | - | 6 - 7 | 697 | 62 | |
BC165701.1 | - | 5 - 6 | 670 | 62 | ||
EH436202.1 | - | 6 - 7 | 723 | 62 | ||
EH462195.1 | - | 1 - 2 | 443 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |