Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amotl2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077606.1 | NP_001071074.1 | 9 - 10 | 2023 | 69 | |
XM_005171208.4 | XP_005171265.1 | 9 - 10 | 2075 | 69 | ||
GenBank mRNA | BC125932.1 | - | 9 - 10 | 2023 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |