Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131860.2 | NP_571935.2 | 6 - 7 | 1238 | 102 | |
GenBank mRNA | BC163403.1 | - | 6 - 7 | 1238 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |