Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hif1al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200405.1 | NP_956699.1 | 12 - 13 | 1632 | 81 | |
XM_005157571.4 | XP_005157628.2 | 12 - 13 | 1702 | 81 | ||
XM_005157572.4 | XP_005157629.2 | 12 - 13 | 1701 | 81 | ||
XM_021481358.1 | XP_021337033.1 | 10 - 11 | 1397 | 81 | ||
GenBank mRNA | AY330295.1 | - | 12 - 13 | 1660 | 81 | |
BC054582.1 | - | 12 - 13 | 1632 | 81 | ||
BC155728.1 | - | 4 - 5 | 377 | 81 | ||
GQ253368.1 | - | 12 - 13 | 1666 | 81 | ||
GQ253369.1 | - | 4 - 5 | 277 | 81 | ||
GQ253370.1 | - | 4 - 5 | 287 | 81 | ||
KR338972.1 | - | 4 - 5 | 363 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |