Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hnrnph1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212589.2 | NP_997754.2 | 8 - 9 | 1067 | 111 | |
XM_021468753.1 | XP_021324428.1 | 7 - 8 | 963 | 111 | ||
GenBank mRNA | BC044161.1 | - | 8 - 9 | 1060 | 111 | |
BC067720.1 | - | 6 - 7 | 788 | 111 | ||
BC164507.1 | - | 7 - 8 | 992 | 111 | ||
CT671502.2 | - | 3 - 4 | 341 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |