Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ube2j1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214767.1 | NP_999932.1 | 7 - 8 | 863 | 93 | |
GenBank mRNA | BC061451.1 | - | 7 - 8 | 863 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |