Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: brf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003536.1 | NP_001003536.1 | 4 - 5 | 652 | 85 | |
XM_009305650.2 | XP_009303925.1 | 3 - 4 | 556 | 85 | ||
GenBank mRNA | BC078247.1 | - | 4 - 5 | 652 | 85 | |
BC154287.1 | - | 4 - 5 | 609 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |