Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: anpepa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044861.1 | NP_001038326.1 | 13 - 14 | 1954 | 131 | |
GenBank mRNA | AW018784.1 | - | 2 - 3 | 146 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |