Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: csnk1g2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045850.2 | NP_001039315.1 | 4 - 5 | 1235 | 72 | |
NM_001290336.1 | NP_001277265.1 | 5 - 6 | 1366 | 72 | ||
XM_005166859.4 | XP_005166916.1 | 5 - 6 | 1400 | 72 | ||
GenBank mRNA | BC122280.1 | - | 4 - 5 | 803 | 72 | |
BC152090.1 | - | 5 - 6 | 1366 | 72 | ||
BC164532.1 | - | 4 - 5 | 710 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |