Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tlr18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001089350.1 | NP_001082819.1 | 3 - 4 | 1106 | 74 | |
XM_005158050.4 | XP_005158107.1 | 3 - 4 | 1357 | 74 | ||
XM_021466668.1 | XP_021322343.1 | 2 - 3 | 1292 | 74 | ||
GenBank mRNA | BC162732.1 | - | 1 - 2 | 973 | 74 | |
BC163840.1 | - | 2 - 3 | 1034 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |