Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pth3r | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131378.1 | NP_571453.1 | 2 - 3 | 462 | 96 | |
XM_021480025.1 | XP_021335700.1 | 2 - 3 | 483 | 96 | ||
GenBank mRNA | AF132085.2 | - | 2 - 3 | 462 | 96 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Danio rerio Probe spans exons |