Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spns1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153663.2 | NP_705949.2 | 5 - 6 | 799 | 86 | |
XM_005163832.4 | XP_005163889.1 | 5 - 6 | 813 | 86 | ||
GenBank mRNA | AF465772.1 | - | 5 - 6 | 755 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Danio rerio Probe spans exons |