Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgef18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214818.1 | NP_999983.1 | 8 - 9 | 1015 | 112 | |
XM_005171284.4 | XP_005171341.1 | 7 - 8 | 1058 | 112 | ||
XM_009298637.3 | XP_009296912.1 | 9 - 10 | 1122 | 112 | ||
XM_021479667.1 | XP_021335342.1 | 8 - 9 | 1095 | 112 | ||
GenBank mRNA | BC070002.1 | - | 8 - 9 | 1015 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |