Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps26bl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002415.2 | NP_001002415.1 | 4 - 5 | 954 | 114 | |
GenBank mRNA | BC076160.1 | - | 4 - 5 | 956 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |