Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: st3gal8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004017.1 | NP_001004017.1 | 3 - 4 | 692 | 95 | |
XM_009306027.3 | XP_009304302.1 | 4 - 5 | 1248 | 95 | ||
XM_017358220.2 | XP_017213709.1 | 3 - 4 | 473 | 95 | ||
GenBank mRNA | AJ705061.1 | - | 2 - 3 | 368 | 95 | |
AJ783740.1 | - | 3 - 4 | 692 | 95 | ||
BC093103.1 | - | 4 - 5 | 1205 | 95 | ||
BC164756.1 | - | 3 - 4 | 723 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |