Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: azin1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007159.2 | NP_001007160.2 | 9 - 10 | 1267 | 144 | |
GenBank mRNA | AB069858.1 | - | 7 - 8 | 901 | 144 | |
BC061960.1 | - | 7 - 8 | 923 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |