Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkbp11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012249.2 | NP_001012249.1 | 5 - 6 | 559 | 96 | |
XM_005165698.4 | XP_005165755.1 | 6 - 7 | 494 | 96 | ||
GenBank mRNA | BC092960.1 | - | 5 - 6 | 464 | 96 | |
BC154504.1 | - | 5 - 6 | 559 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |