Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cfap53 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045130.2 | NP_001038595.2 | 1 - 2 | 172 | 100 | |
GenBank mRNA | BC124260.1 | - | 1 - 2 | 172 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |