Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: g6pc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080066.1 | NP_001073535.1 | 2 - 3 | 432 | 80 | |
GenBank mRNA | BC128866.1 | - | 2 - 3 | 432 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |