Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kdm6al | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080193.1 | NP_001073662.1 | 9 - 10 | 992 | 103 | |
XM_005168796.4 | XP_005168853.1 | 9 - 10 | 3693 | 103 | ||
GenBank mRNA | BC129198.1 | - | 9 - 10 | 992 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |