Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hspa4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199857.1 | NP_956151.1 | 12 - 13 | 1679 | 72 | |
GenBank mRNA | BC048063.1 | - | 12 - 13 | 1679 | 72 | |
BC065970.1 | - | 12 - 13 | 1708 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |