Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chek1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200193.1 | NP_956487.1 | 9 - 10 | 1143 | 112 | |
XM_021468776.1 | XP_021324451.1 | 10 - 11 | 1265 | 112 | ||
GenBank mRNA | BC045904.1 | - | 9 - 10 | 1143 | 112 | |
BC164512.1 | - | 8 - 9 | 940 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |