Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec22ba | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213384.1 | NP_998549.1 | 3 - 4 | 531 | 105 | |
GenBank mRNA | BC046043.1 | - | 3 - 4 | 531 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |