Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hormad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002357.1 | NP_001002357.1 | 12 - 13 | 1014 | 85 | |
XM_021466675.1 | XP_021322350.1 | 10 - 11 | 1621 | 85 | ||
GenBank mRNA | BC075898.1 | - | 12 - 13 | 1014 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |