Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rhocb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002445.1 | NP_001002445.1 | 2 - 3 | 222 | 80 | |
GenBank mRNA | AY865557.1 | - | 1 - 2 | 159 | 80 | |
BC076026.1 | - | 2 - 3 | 222 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |