Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ela3l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024408.1 | NP_001019579.1 | 5 - 6 | 496 | 144 | |
XM_005171867.4 | XP_005171924.1 | 5 - 6 | 564 | 144 | ||
XM_021476165.1 | XP_021331840.1 | 5 - 6 | 564 | 144 | ||
GenBank mRNA | AY583322.1 | - | 5 - 6 | 492 | 144 | |
BC095070.1 | - | 5 - 6 | 496 | 144 | ||
BC165005.1 | - | 3 - 4 | 319 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |